Order Entry
Switzerland
ContactUsLinkComponent
Aniline-[ring-D5] 2000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Aniline-[ring-D5] 2000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Catalog # EHERYA10262600MB
Supplier:  EHRENSTORFER
CAS Number:  
Aniline-[ring-D5] 2000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Catalog # EHERYA10262600MB
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    4165-61-1
  • Description:
    Aniline-[ring-D5] 2,000 µg/ml in tert-butyl methyl ether
  • Formula:
    C₆₂H₅H₂N
  • Description:
    Aniline-[ring-D5]

Specifications

About this item