D(-)-Luciferin
Glass bottle for solids, 100 mg
Catalog # MSPP102111
Marketsource
Supplier: MARKET SOURCE PART PROCESS
D(-)-Luciferin
Catalog # MSPP102111
Supplier: MARKET SOURCE PART PROCESS
Marketsource
CAS Number: 2591-17-5
Jump to:
Product Details & Documents
Synonyms: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
- CAS Number:2591-17-5
- Formula:C₁₁H₈N₂O₃S₂
- MDL Number:MFCD00042929
- Molecular Weight:280.3228 g/mol
- EINECS:219-981-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



