Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, TRIS solution pH 7,5 (2 mol/L)

Buffer, TRIS solution pH 7,5 (2 mol/L)

TRIS buffer solution pH 7,5 (2 mol/l), 1 KIT
Catalog # EURLEMR253001BK
Supplier:   EUROCLONE
Buffer, TRIS solution pH 7,5 (2 mol/L)
Catalog # EURLEMR253001BK
Supplier:  EUROCLONE
CAS Number:  
Buffer, TRIS solution pH 7,5 (2 mol/L)

TRIS buffer solution pH 7,5 (2 mol/l), 1 KIT

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    TRIS buffer solution pH 7,5 (2 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.