Buffer, TRIS solution pH 7,4 (0,1 mol/L), Millipore®
TRIS buffer solution pH 7,4 (0,1 mol/l), Glass bottle, 100 mL
Supplier: Merck Millipore (Calbiochem)
Buffer, TRIS solution pH 7,4 (0,1 mol/L), Millipore®
Catalog # 648315-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Jump to:
Product Details & Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Description:TRIS buffer solution pH 7,4 (0,1 mol/l)



