Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, bicarbonate, 1M solution, pH 9,0

Buffer, bicarbonate, 1M solution, pH 9,0

Bicarbonate, 1M buffer solution, pH 9,0, Glass bottle, 250 mL
Catalog # J60116.AK
Buffer, bicarbonate, 1M solution, pH 9,0
Catalog # J60116.AK
CAS Number:  
Buffer, bicarbonate, 1M solution, pH 9,0
Buffer, bicarbonate, 1M solution, pH 9,0

Bicarbonate, 1M buffer solution, pH 9,0, Glass bottle, 250 mL

CAS Number:  
Price
SFr. 101.00
Log in for availability
1 * 250 mL
Product Details & Documents

CAS No.: 144-55-8
Physical form: liquid
MDL No.: MFCD00003528

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Description:
    Bicarbonate, 1M buffer solution, pH 9,0
  • Cat. No.:
    J60116.AK
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00003528
  • CAS No.:
    144-55-8

Specifications