SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organic Standards
p-Terphenyl-[D14] 200 µg/ml in n-nonane (98% D), CIL
p-Terphenyl-[D14] 200 µg/ml in n-nonane (98% D), CIL
p-Terphenyl-[D14] 200 µg/ml in n-nonane (98% D), CIL
PROOCILDLM-382-1.2
:
LGC Standards PROMOCHEM
:
p-Terphenyl-[D14] 200 µg/ml in n-nonane (98% D), CIL
PROOCILDLM-382-1.2
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,2 mL
CAS No.:
1718-51-0
Description:
p-Terphenyl-[D14] 200 µg/ml in n-nonane (98% D)
Formula:
C₁₈D₁₄
Description:
p-Terphenyl-[D14]