Pesticide reference standard, Carbosulfan 100 µg/ml in isooctane, Dr Ehrenstorfer
EHERXA11030000IO
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:55285-14-8
- Description:Carbosulfan 100 µg/ml in isooctane
- Formula:C₂₀H₃₂N₂O₃S
- Description:Carbosulfan