Erlenmeyer flasks, with PP screw cap, Azlon®
PMP, transparent.
- Good chemical resistance
- Heat resistant up to 150 °C (up to 170 °C for short periods)
- Flask and cap can be autoclaved at 121 °C
With red, easy to read graduations. Wide mouth with screw cap GL 40 or GL 52, can also be closed using standard ground PP stoppers.
Frequently Bought Together