Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cutting board

Cutting board

Cutting board, PP, White, 300×210×9 mm
Catalog # 216-1300
Supplier:   SciLabware
Cutting board
Catalog # 216-1300
Supplier:  SciLabware
Cutting board
Cutting board

Cutting board, PP, White, 300×210×9 mm

Price
SFr. 67.10
Log in for availability
1 * 1 ST
Product Details & Documents

Durable cutting board made of PP.

  • Autoclavable at 121 °C
  • Good chemical resistance
  • Unbreakable in normal use
  • Non absorbent and easy to clean

Documents

Specifications
  • Colour:
    White
  • Description:
    Cutting board, PP
  • W×D×H:
    300×210×9 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.