Order Entry
Switzerland
ContactUsLinkComponent
Mag-Bind® Bacterial DNA 96 Kits
Mag-Bind® Bacterial DNA 96 Kits
Supplier:  OMEGA BIO-TEK
Certificates
Mag-Bind® Bacterial DNA 96 Kits
Supplier:  OMEGA BIO-TEK

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Bacterial DNA
  • Sample type:
    Swabs
    Bacteria
    Biofluids (serum, plasma, saliva)
  • Target molecule:
    DNA
  • Sample size:
    Up to 0,5 ml
  • Description:
    Mag-Bind® Bacterial DNA 96 Kit

Specifications

Related Information

Frequently Bought Together