Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ion selective electrode, digital, NitraLyt® Plus

Ion selective electrode, digital, NitraLyt® Plus

Digital sensor for nitrate measurement, NitraLyt® Plus 700 IQ, 1 - 1000 mg/l (NO₃-N); 5 - 4500 mg/l (NO₃⁻); 1 - 1000 mg/l (Cl⁻), 4 - 11, 0...+40 °C, Stainless steel, Screw/plug connector for SACIQ sensor connection cable, 40×392 mm, Wastewater measurements
Catalog # WTWO107080
Supplier:   WTW ONLINE
Ion selective electrode, digital, NitraLyt® Plus
Catalog # WTWO107080
Supplier:  WTW ONLINE
Ion selective electrode, digital, NitraLyt® Plus
Ion selective electrode, digital, NitraLyt® Plus

Digital sensor for nitrate measurement, NitraLyt® Plus 700 IQ, 1 - 1000 mg/l (NO₃-N); 5 - 4500 mg/l (NO₃⁻); 1 - 1000 mg/l (Cl⁻), 4 - 11, 0...+40 °C, Stainless steel, Screw/plug connector for SACIQ sensor connection cable, 40×392 mm, Wastewater measurements

Price
SFr. 6’160.00
Log in for availability
1 * 1 ST
Product Details & Documents

With the NitraLyt® Plus 700 IQ, nitrate can be determined directly in the medium. This makes measured values available in a timely manner and they can easily be used for regulation purposes.

  • Calibration free, long-term stability
  • Economical and cost efficient

Measuring range or resolution (NO₃-N): 1 - 1000 mg/l (1 mg/l); 0,1 - 100 mg/l (0,1 mg/l)

Specifications
  • Concentration:
    1 - 1000 mg/l (NO₃-N); 5 - 4500 mg/l (NO₃⁻); 1 - 1000 mg/l (Cl⁻)
  • Connection:
    Screw/plug connector for SACIQ sensor connection cable
  • Ø×L:
    40×392 mm
  • For:
    Wastewater measurements
  • Description:
    Digital sensor for nitrate measurement, NitraLyt® Plus 700 IQ
  • Material:
    Stainless steel
  • Temp.:
    0...+40 °C
  • pH:
    4 - 11
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Chloride compensation electrode, VARiON® Plus Cl, meausring range 1 - 1000 mg/l Cl-
    Chloride compensation electrode, VARiON® Plus Cl, meausring range 1 - 1000 mg/l Cl-
    WTWO107047
    VARiON® Plus 700 IQ and NitraLyt® Plus
    Log in for availability
    1 * 1 ST - SFr. 550.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.