Media bottles, graduated, with screw cap
Supplier: BELLCO GLASS
Certificates
About this item
Manufactured from type 1, class B borosilicate glass.
- White graduations
- Marking spot
Supplied with a 33-430 nontoxic, rubber-lined black phenolic cap Large opening and sloping shoulders facilitate pouring and cleaning.