Order Entry
Switzerland
ContactUsLinkComponent
Aerosol filter holders
Aerosol filter holders
Supplier:  Merck
Certificates
undefined
Aerosol filter holders
Supplier:  Merck

Specifications

  • Catalog No:
  • MILFXX5004700
  • MILFXX5004740
  • MILFXX5004710
  • Filtration area:
  • 9,6 cm²
  • 9,6 cm²
  • 9,6 cm²
  • Type:
  • Aerosol
  • Aerosol
  • Aerosol
  • Dimensions:
  • Length: 178 mm, Ø: 69 mm
  • Length: 102 mm, Ø: 69 mm
  • Length: 102 mm, Ø: 69 mm
  • Inlet/outlet connections:
  • Hose connector (11 mm O.D.), threaded for flow-limiting orifices
  • Hose connector (11 mm O.D.), threaded for flow-limiting orifices
  • Hose connector (11 mm O.D.), threaded for flow-limiting orifices
  • Material:
  • Stainless steel
  • Stainless steel
  • Stainless steel

Specifications

Frequently Bought Together