ContactUsLinkComponent
Sampling fork, Corning® Gosselin™
Sampling fork, Corning® Gosselin™
Catalog # 215-2296
Supplier:  Corning
   Low Manufacturing Impact
undefined
Sampling fork, Corning® Gosselin™
Catalog # 215-2296
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Colour:
    White
  • Description:
    Sampling fork, individually packed
  • Length:
    155 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Borre
  • Pk:
    1.000
Specifications
About this item
PS, white.
  • Food contact approved: Raw material is compliant with regulation EC No. 1935/2004
  • Ideal for dairy industries
  • Sterile
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904