VWR® Q Path® Formalin Neutraliser Safety Pads
Supplier: VWR Collection
Jump to:
About this item
Absorbs the odours (neutralises formalin). Ensures good contrast for biopsies.
- Different sizes available
- Very useful for the bottom of transport boxes and small formalin spills
Protects knives and cutting boards against premature wear.
3 Options Available Below
Product Details & Documents
Absorbs the odours (neutralises formalin). Ensures good contrast for biopsies.
- Different sizes available
- Very useful for the bottom of transport boxes and small formalin spills
Protects knives and cutting boards against premature wear.
Documents
Product Family Options
Product Information