Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Evacuation body splint

Evacuation body splint

Evacuation body splint, PP bed with nylon straps (44 mm), 2 m
Catalog # 111-1387
Supplier:   HONEYWELL SAFETY
Evacuation body splint
Catalog # 111-1387
Supplier:  HONEYWELL SAFETY
Evacuation body splint
Evacuation body splint

Evacuation body splint, PP bed with nylon straps (44 mm), 2 m

Price
SFr. 1’090.00
Log in for availability
1 * 1 ST
Product Details & Documents

Lightweight body splint that is ideal for confined space evacuation. Once wrapped around a casualty, it provides a rigid splint. Ideal for the army and fire brigades.

  • Padded shoulder straps provide extra support and protection for the casualty
  • Fitted with foot supports, it can be lifted vertically
  • Can be stored in a compact bag and is easy to transport
Specifications
  • Description:
    Evacuation body splint, PP bed with nylon straps (44 mm)
  • Length:
    2 m
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.