ContactUsLinkComponent
Scaffolding lanyards, Barracuda™, Miller®
Scaffolding lanyards, Barracuda™, Miller®
Supplier:  HONEYWELL SAFETY
Certificates
undefined
Scaffolding lanyards, Barracuda™, Miller®
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • 111-1390
  • 111-1392
  • Model:
  • 1,2 m
  • 2 m
  • Breaking strength:
  • > 2200 daN
  • > 2200 daN
  • Connector type:
  • Zinc finished steel
  • Galvanized steel
  • Weight:
  • 1,22 kg
  • 1,20 kg