Chemical Resistant Overalls, 4515
About this item
These disposable overalls made of highly breathable SMS material provide basic protection against dry particles, certain liquid chemical splashes and hazardous dusts such as asbestos fibres.
- Breathable material technology for reduced heat stress
- Robust, low linting
- Two-way zip with storm flap (incl. tape tabs) for convenience and additional protection
- Elasticated cuffs, waist and ankles for a better fit and freedom of movement
Typical light duty applications may include asbestos inspection, insulation laying, cement manufacturing and metal grinding and polishing.
Certifications: Type 5/6, EN 1073-2; Cat. III
20 Options Available Below
Product Details & Documents
These disposable overalls made of highly breathable SMS material provide basic protection against dry particles, certain liquid chemical splashes and hazardous dusts such as asbestos fibres.
- Breathable material technology for reduced heat stress
- Robust, low linting
- Two-way zip with storm flap (incl. tape tabs) for convenience and additional protection
- Elasticated cuffs, waist and ankles for a better fit and freedom of movement
Typical light duty applications may include asbestos inspection, insulation laying, cement manufacturing and metal grinding and polishing.
Certifications: Type 5/6, EN 1073-2; Cat. III
Specifications for Product Family
Specifications
- Catalog No:
- 113-6259
- 113-6258
- 113-6257
- 113-6260
- 113-6254
- 113-6255
- 113-6256
- 113-6265
- 113-6264
- 113-6263
- 113-6266
- 113-6261
- 113-6262
- 113-6279
- 113-6278
- 113-6277
- 113-6280
- MMMA4515WXXL
- 113-6275
- 113-6276
- Model:
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- 4515
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Garment application:
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
- Chemical protection|Laboratory|Industrial
Specifications
Product Family Options
Product Information
- ColourClothing SizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-6259Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6258Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6257Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6260Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6254Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6255Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6256Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6265Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6264Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6263Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6266Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6261Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6262Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6279Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6278Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6277Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6280Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.MMMA4515WXXLModel4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6275Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial - Specifications:
More
Specifications
Cat. No.113-6276Model4515Sleeve or leg lengthLongGarment applicationChemical protection|Laboratory|Industrial