Orders LinkContactUsLinkComponent
Universal absorbent pillows
Universal absorbent pillows
Supplier:   asecos
Universal absorbent pillows
Supplier:  asecos
Certificates
Universal absorbent pillows
Universal absorbent pillows
2 Options Available Below
Product Details & Documents
Universal pillows for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Optimal for those hard to reach places
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
Product Family Options
Product Information
  • Packed
    L×W
    Availability
    Price
  • MSGA9943
    Universal pillow
    Universal pillow
    10 pillows/pack
    460×460 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 150.00
    SFr. 150.00
    1 * 1 ST
       SFr. 150.00
  • MSGA9944
    Universal pillow
    Universal pillow
    20 pillows/pack
    250×250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 184.00
    SFr. 184.00
    1 * 1 ST
       SFr. 184.00