Order Entry
Switzerland
ContactUsLinkComponent
 
Affinity chromatography media, Ni Sepharose™ High Performance
 :  Cytiva
 
undefined
Affinity chromatography media, Ni Sepharose™ High Performance
 :  Cytiva
 

 

  • pH range:
    3 - 12
  • Matrix:
    Highly cross-linked agarose, 6%
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    At least 40 mg histidine-tagged protein/ml
  • Total ionic capacity:
    15 µmol Ni2+/ml

 

Frequently Bought Together