Order Entry
Switzerland
ContactUsLinkComponent
Protein A column, HiScreen™ MabSelect SuRe™
Protein A column, HiScreen™ MabSelect SuRe™
  28-9269-77
 :  Cytiva
undefined
Protein A column, HiScreen™ MabSelect SuRe™
  28-9269-77
 :  Cytiva

 

  • Particle Size:
    85 µm
  • Max. pressure:
    3 bar
  • Capacity:
    4,7 ml
  • Description:
    HiScreen™ MabSelect SuRe™
  • Seq:
    01
  • pH range:
    3 - 12
  • Storage:
    2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
  • Column hardware:
    PP
  • Bed dimensions:
    7,7×100 mm
  • Flow velocity:
    <500 cm/h
  • Pk:
    1

 

 

The HiScreen MabSelect SuRe column uses protein A chromatography for antibody purification. This protein A column is prepacked with MabSelect SuRe affinity resin and is part of our process development platform. These packed columns are used for method optimization and parameter screening for capture of monoclonal antibodies.

  • The novel, alkali-tolerant recombinant protein A ligand of MabSelect Sure allows cleaning-in-place (CIP)
  • Prepacked, ready-to-use columns for convenient process development
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography resin using the same linear fluid velocity

The small column volume of 4,7 ml and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up of the protein purification process. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.