Order Entry
Switzerland
ContactUsLinkComponent
Cation exchange chromatography columns, RESOURCE™ S
Cation exchange chromatography columns, RESOURCE™ S
Supplier:  Cytiva
Certificates
undefined
Cation exchange chromatography columns, RESOURCE™ S
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 17-1180-01
  • 17-1178-01
  • Description:
  • RESOURCE™ S
  • RESOURCE™ S
  • Max. pressure:
  • 6 bar
  • 15 bar
  • pH range:
  • 2 - 13
  • 2 - 13
  • Storage:
  • 4 to 30 °C, 20% Ethanol, 0,2 M Sodium Acetate
  • 4 to 30 °C, 20% Ethanol, 0,2 M Sodium Acetate

Specifications

Frequently Bought Together