Informazioni su questo articolo
Mono-sized 15 um rigid beads gives low back pressure and high flow rates allowing for high productivity.
- Polymeric, strong anion exchanger designed for high resolution polishing purification of proteins, peptides and oligonucleotides
- High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH
- Bioprocessing medium supported for industrial applications
- Particle structure is consistent both bead-to-bead and batch-to batch
SOURCE 15Q is a synthetic high performance, preparative, chromatography medium, based on a 15 µm monosized, rigid polystyrene/divinyl benzene polymer matrix. It is modified with quaternary ammonium (Q) strong anion exchange groups. SOURCE media have excellent physical and chemical characteristics, allowing high flow rates and consistent performance.
Through the combination of high quality assurance standards and a patented manufacturing process, particle structure is consistent both bead-to-bead and batch-to batch. With its very high resolution, SOURCE 15Q is suitable for polishing steps in in both research and industrial applications. As member of the BioProcess media range, SOURCE 15Q meets industrial demands with security of supply and comprehensive technical and regulatory support.
Strong anion exchanger designed for high resolution polishing purification of proteins, peptides and oligonucleotides.
Specifiche
- pH range:2 - 12
- Matrix:Polystyrene/divinylbenzene
- Ligand:Quaternary ammonium
- Storage temperature:4 - 30 °C
- Ligand coupling method:Long, hydrophilic spacer arms
Specifiche
Informazioni correlate
17094701_DATA FILE 17094720_DATA FILE
Scheda dati
17094701_HAND BOOK 17094701_HAND BOOK 1 17094701_INSTRUCTIONS 17094701_INSTRUCTIONS 2 17094701_INSTRUCTIONS 1
Manuale
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements