Order Entry
Switzerland
ContactUsLinkComponent
Affinity chromatography media, MabSelect SuRe™ LX
Affinity chromatography media, MabSelect SuRe™ LX
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography media, MabSelect SuRe™ LX
Supplier:  Cytiva

Specifications

  • Matrix:
    Highly cross-linked agarose
  • Ligand:
    Alkali-stabilized Protein A-derived (E. coli)
  • Storage temperature:
    2 - 8 °C
  • Binding capacity:
    60 mg human IgG/ml
  • Flow rate:
    500 cm/h

Specifications

Related Information

Frequently Bought Together