Order Entry
Switzerland
ContactUsLinkComponent
 
Hydrophobic interaction chromatography media, Capto™ Butyl
Catalog # 17-5459-01
Supplier:  Cytiva
CAS Number:  
undefined
Hydrophobic interaction chromatography media, Capto™ Butyl
Catalog # 17-5459-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    75 µm
  • Pk:
    25 ml
  • Description:
    Capto™ Butyl
  • Seq:
    01
  • Environmentally Preferable:
  • pH range:
    3 - 13
  • Matrix:
    Highly cross-linked agarose
  • Ligand:
    Butyl
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    27 mg BSA/ml
  • Type:
    Liquid

Specifications

About this item

Capto™ Butyl is used in the capture and intermediate stages of protein purification. The combination of high capacity, high flow rate and low back pressure reduces process cycle times.

  • Improved productivity and process economy in downstream operations
  • Very high flow rates and large sample volume processing
  • Excellent chemical stability