Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Digital Timer, One Channel
VWR®, Digital Timer, One Channel
Catalog # 609-0190
Supplier:  VWR Collection
undefined
VWR®, Digital Timer, One Channel
Catalog # 609-0190
Supplier:  VWR Collection

Specifications

  • Colour:
    White
  • Description:
    Digital timer, single channel
  • W×D×H:
    64×15,2×64 mm
  • Accuracy:
    ±5 s/day
  • Pk:
    1

Specifications

About this item

This single-channel timer is switchable between 19 hours, 59 minutes and 99 minutes, 59 seconds, and has a 3-key operation for ease of use.

  • Count direction: Up/down
  • Last time set memory
  • Audible alarm; displays elapsed time after countdown
Certifications: Certificate of calibration and individual serial number, these electronic timers are tested and calibrated in an exclusive triple-accredited/-registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery: Supplied with fold-away stand, pocket clip, magnetic back, one AAA battery and certificate of calibration. Includes a statement of traceability indicating accuracy to NIST.