Order Entry
Switzerland
ContactUsLinkComponent
 
Ion exchange chromatography media, DEAE Sephacel™
Catalog # 17-0500-01
Supplier:  Cytiva
CAS Number:  
undefined
Ion exchange chromatography media, DEAE Sephacel™
Catalog # 17-0500-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    ~100 µm
  • Pk:
    500 mL
  • Description:
    DEAE Sephacel™
  • Environmentally Preferable:
  • pH range:
    2 - 12
  • Storage temperature:
    4 - 8 °C
  • Binding capacity:
    10 mg Thyroglobulin (MW 669 000)/ml drained resin, 160 mg HSA (MW 68 000)/ml drained resin
  • Total ionic capacity:
    0,10 - 0,14 mmol Cl-/ml
  • Flow rate:
    <50 cm/h
  • Exclusion limit:
    1000000 daltons
  • Type:
    Liquid

Specifications

About this item

DEAE Sephacel™ is a weak anion exchanger based on beaded cellulose. The ion exchange group is diethylaminoethyl, which remains charged and maintains consistently high capacity over the entire working range. This media is macroporous and has an exclusion limit of ~10000 kDa for globular proteins. Supplied pre-swollen in 20% ethanol.

The ion exchange group is diethylaminoethyl, which remains charged and maintains consistently high capacity over the entire working range. This media is macroporous and has an exclusion limit of approximately 1000000 daltons for globular proteins. Supplied pre-swollen in 20% ethanol. Autoclavable in 0.1 M NaCl at 121 °C for 30 min. Avoid using oxidizing materials. Stable in aqueous buffers, 1.0 M acetic acid, 1.0 M NaOH, 8 M Urea, 8 M guanidine hydrochloride, ethanol, methanol. Negligible volume variation due to changes in pH or ionic strength.