ContactUsLinkComponent
2-D Protein Extraction Buffer
2-D Protein Extraction Buffer
Catalog # 28-9435-22
Supplier:  Cytiva
CAS Number:  
undefined
2-D Protein Extraction Buffer
Catalog # 28-9435-22
Supplier:  Cytiva
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    2-D Protein Extraction Buffer trial kit, 6×10 ml
  • Seq:
    07
Specifications
About this item
Efficient cell lysis and protein extraction are key steps for achieving high-quality results in downstream applications. The 2-D protein extraction buffer trial kit is designed for those who experiment for establishing a suitable extraction and solubilization buffer for their protein samples.
  • Convenient way to prepare high-quality protein lysates
  • Designed to produce high spot resolution for 2-D gel analysis
These 2-D Protein Extraction Buffer is modifications of protein solubilization and IPG-strip rehydration buffers designed to produce better spot resolution for 2-D gel analysis.well-studied.
Six buffers are available as a dry powder, which is rehydrated with the provided diluent. The optimal buffer will depend on the nature of the sample, and the 2-D protein extraction buffer trial kit allows to evaluate each extraction buffer to find the most suitable buffer for the sample. extraction buffer-I and -II are suitable for most applications. However, for stronger solubilization effects, the Extraction Buffer-III, -IV, -V or -VI may be used.
Delivery information: The trial kit consists of one each of the 2-D protein extraction buffers -I to -VI with their corresponding diluents. Instructions supplied with the buffer.