Order Entry
Switzerland
ContactUsLinkComponent
Affinity chromatography columns, HiTrap™ MabSelect SuRe™
Affinity chromatography columns, HiTrap™ MabSelect SuRe™
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography columns, HiTrap™ MabSelect SuRe™
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 11-0034-93
  • 11-0034-94
  • 29-0491-04
  • 11-0034-95
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Storage:
  • 2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
  • 2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
  • 2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
  • 2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)

Specifications

Frequently Bought Together