Platinum electrode
BTXI45-0510
: BTX
- Application:In-Vivo/Ex-Vivo tissues gene or drug delivery
- Description:Platinum needle electrode kit, L-shaped, with cables
- Pk:1 KIT
This needle style platinum electrode is specifically designed for in vivo applications on the most fragile of tissue types, such as brain tissue. The ultra thin electrode enables pinpoint transfection for greater ease and efficiency in fragile or in accessible tissue.
- Ideal for delivering the electrical pulses directly to oocytes or embryos for nuclear transfer fusion applications
- L-shaped electrodes have 3 mm tip length in order to accommodate the most research needs in small animal models
Generator compatibility: ECM 830 and ECM 2001
Voltage range: 0 to 100 V
Pulse length range: 10 µsec to 100 msec
Needle length: 3 mm
Electrode length: 3 mm