Order Entry
Switzerland
ContactUsLinkComponent
PIG® Overpack Dolly
PIG® Overpack Dolly
Catalog # NEWPPAK414
Supplier:  NEW PIG
undefined
PIG® Overpack Dolly
Catalog # NEWPPAK414
Supplier:  NEW PIG

Specifications

  • Ø×H:
    690×170 mm
  • Colour:
    Black
  • Description:
    Overpack dolly
  • Pk:
    1
  • Weight:
    12,7 kg

Specifications

About this item

The wide-base design helps ensure stability during transport of overpacks or rapid response with a PIG overpack spill kit.

  • Heavy-duty rolled steel construction holds up to 363 kg
  • Lockable casters keep overpack stationary, even on uneven surfaces