2-Methoxy-3-pyridineboronic acid
Catalog # ACTIAS50873G5
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:152.95 g/mol
CAS Number:163105-90-6
Formula:C₆H₈BNO₃
MDL Number:MFCD03411572