Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Azure A
Azure A
Azure A
:
Thermo Fisher Scientific
Azure A
:
Thermo Fisher Scientific
:
531-53-3
:
:
C₁₄H₁₄ClN₃S
:
MFCD00012112
:
291.797 g/mol
:
14,927
:
290 °C (dec.)
:
208-510-7
:
GHS07
:
Ambient
msds_ALFAJ61346
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Standard Line Transfer Pipettes
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Blender Bags
: VWR Collection
Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
: Merck
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol, LiChrosolv®, gradient grade for liquid chromatography, Supelco®
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
: VWR Chemicals
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF