5-Bromo-2-(trifluoromethyl)pyridine
Supplier: ACTIVATE SCIENTIFIC
Certificates
About this item
Synonyms: 5-Bromo-2-trifluoromethylpyridine
CAS Number:436799-32-5
:
Formula:C₆H₃BrF₃N
MDL Number:MFCD06657686
:226.00 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together