8-Methoxyisoquinoline
Glass bottle for solids, 5 g
Catalog # ACTIAS11337G5
Marketsource
Supplier: ACTIVATE SCIENTIFIC
8-Methoxyisoquinoline
Catalog # ACTIAS11337G5
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 1723-70-2
Jump to:
Product Details & Documents
- CAS Number:1723-70-2
- Formula:C₁₀H₉NO
- MDL Number:MFCD09258766
- Molecular Weight:159.190 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



