Dextran, M.W. ~ 40,000
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ampicillin sodium salt 91.0-102.0%, BioChemica®
: PanReac AppliChem
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals