Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
di-Lithium tetraborate, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
  4503-01
 :  Avantor
 :  
undefined
 
 
 
di-Lithium tetraborate, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
  4503-01
 :  Avantor
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • For Laboratory,Research,or Manufacturing Use:
    1.95 - 2.05
  • B₂O₃/Li₂O Ratio:
    1.95 - 2.05
  • Loss on Drying at 285°C:
    ≤ 1 %
  • Lithium Oxide (Li2O):
    ≥ 17.0 %
  • Water (H₂O):
    ≤ 1.0 %
  • Ferric Oxide (Fe2O3):
    ≤ 0.03 %
  • Sodium (Na):
    ≤ 0.015 %
  • Calcium (Ca):
    ≤ 0.015 %
  • Silicon Dioxide (SiO2):
    ≤ 0.03 %
  • Titanium Dioxide (TiO2):
    ≤ 0.02 %

 

 

 :
 :169.12 g/mol
 :14,05524
 :918 °C
 :1.4 g/cm³ (25 °C)
 :234-514-3
 :GHS05
 :GHS08
 :GHS07
 :Ambient
 :12007-60-2
 :Li₂B₄O₇
 :MFCD00011083