Ethyl-5-bromo-2-chlorobenzoate
Catalog # ACTIAS23637G25
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:263.52 g/mol
CAS Number:76008-73-6
Formula:C₉H₈BrClO₂
MDL Number:MFCD00013635