Order Entry
Switzerland
Orders LinkContactUsLinkComponent
2,6-Dichlorophenethylamine

2,6-Dichlorophenethylamine

Marketsource
Supplier:   ACTIVATE SCIENTIFIC
2,6-Dichlorophenethylamine
Marketsource
Certificates
2,6-Dichlorophenethylamine

3 Options Available Below

Product Details & Documents
Synonyms:  2-(2-Aminoethyl)-1,3-dichlorobenzene, 1-Amino-2-(2,6-dichlorophenyl)ethane, 2-(2,6-Dichlorophenyl)ethylamine, 2,6-Dichlorophenethylamine
  • CAS Number:14573-23-0
  • Formula:C₈H₉Cl₂N
  • MDL Number:MFCD00060612
  • Molecular Weight:190.07 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource