Phenol 99% unstabilised, Ultrapure
:108-95-2
:
:C₆H₅OH
:MFCD00002143
:94.11 g/mol
:14,7241
:181 °C
:40…43 °C
:1.071
:79 °C (175 °F)
:Ambient
:203-632-7
:1671
:6.1,II
:01-2119471329-32
:GHS06
:GHS05
:GHS08
Frequently Bought Together