2-Bromophenol
Supplier: ACTIVATE SCIENTIFIC
Certificates
About this item
CAS Number:95-56-7
:
Formula:C₆H₅BrO
MDL Number:MFCD00002146
:173.01 g/mol
Merck Index:13,01413
Boiling Pt:195 °C (1013 hPa)
Melting Pt:5.6 °C
Density:1.49 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-432-7
UN:3082
ADR:9,III
Frequently Bought Together