2-Hydroxy-3-(trifluoromethyl)pyridine
Catalog # ACTIAS4023G5
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:163.1 g/mol
CAS Number:22245-83-6
Formula:C₆H₄F₃NO
MDL Number:MFCD00153194