1-(4-Nitrophenyl)imidazole
Catalog # ACTIAS30297G5
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:189.17 g/mol
CAS Number:2301-25-9
Formula:C₉H₇N₃O₂
MDL Number:MFCD00060491