Order Entry
Switzerland
ContactUsLinkComponent
 
Zinc, granules, BAKER ANALYZED® ACS, J.T.Baker®
  4252-01
 :  Avantor
 :  
undefined
Zinc, granules, BAKER ANALYZED® ACS, J.T.Baker®
  4252-01
 :  Avantor
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • For Laboratory,Research,or Manufacturing Use:
    = 99.8 %
  • Assay (Zn) (by EDTA titrn):
    ≥99.8 %
  • Iron (Fe):
    ≤0.01 %
  • Lead (Pb):
    ≤0.01 %
  • Suitability for Arsenic Determination:
    Passes Test
  • Trace Impurities - Arsenic (As):
    ≤0.100 ppm
  • Mesh - On U.S. No. 40 Sieve:
    ≤5 %
  • Mesh - Thru U.S. No. 70 Sieve:
    ≤30 %

 

 

 :
 :65.39 g/mol
 :12,10255
 :908 °C (1013 hPa)
 :420 °C
 :7.14 g/cm³ (20 °C)
 :Ambient
 :231-175-3
 :7440-66-6
 :Zn
 :MFCD00011291