2-Chloro-4-(trifluoromethyl)pyridine
Catalog # ACTIAS7182G5
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:181.54 g/mol
Boiling Pt:146…147 °C (1013 hPa)
Melting Pt:–1 °C
Density:1.411 g/cm³ (20 °C)
UN:1992
ADR:3,III
CAS Number:81565-18-6
Formula:C₆H₃ClF₃N
MDL Number:MFCD00042224