5-(Trifluormethyl)-m-anisidin
Catalog # ACTIAS1596G5
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Packung:Glass bottle for solids
- VE:5 g
- Seq:3173
Specifications
About this item
:
:191.15 g/mol
EINECS:206-487-8
UN:2811
ADR:6.1,III
CAS Number:349-55-3
Formula:C₈H₈F₃NO
MDL Number:MFCD00000393