Order Entry
Switzerland
ContactUsLinkComponent
3-Fluoro-p-anisidine
3-Fluoro-p-anisidine
Catalog # ACTIAS31263G5
CAS Number:  
3-Fluoro-p-anisidine
Catalog # ACTIAS31263G5
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :141.15 g/mol
UN:2811
ADR:6.1,III
CAS Number:366-99-4
Formula:C₇H₈FNO
MDL Number:MFCD00075040