3-Fluoropyrocatechol
Catalog # ACTIAS33505G5
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:128.1 g/mol
Boiling Pt:218 °C (1013 hPa)
Boiling Pt:218 °C (1013 hPa)
Melting Pt:71 °C
Density:1,514 g/cm³ (20 °C)
CAS Number:363-52-0
Formula:C₆H₅FO₂
MDL Number:MFCD00042582