Rifampicin for molecular biology
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13292-46-1
:
Formula:C₄₃H₅₈N₄O₁₂
MDL Number:MFCD00151389
:822.95 g/mol
Merck Index:14,8216
Melting Pt:183…188 °C
EINECS:236-312-0
:GHS07
Storage Temperature:Freezer
Related Information
Frequently Bought Together