Ethyl-4-cyanobenzoate
Supplier: ACTIVATE SCIENTIFIC
Certificates
About this item
CAS Number:7153-22-2
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD00001824
:175.19 g/mol
Boiling Pt:152 °C (14 mmHg)
Melting Pt:54 °C
EINECS:230-500-6
Frequently Bought Together